Assay Details
Gene Symbol: LOC101707360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850338.2 | XP_004850395.1 | 7 - 8 | 706 | 71 | |
XM_021262850.1 | XP_021118509.1 | 7 - 8 | 782 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
