Assay Details
| Gene Symbol: Eloc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842049.3 | XP_004842106.1 | 2 - 3 | 190 | 74 | |
| XM_021256013.1 | XP_021111672.1 | 3 - 4 | 207 | 74 | ||
| XM_021256014.1 | XP_021111673.1 | 2 - 3 | 356 | 74 | ||
| XM_021256015.1 | XP_021111674.1 | 2 - 3 | 72 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004842049.3, XM_021256013.1, XM_021256014.1, XM_021256015.1 transcripts.
Set Membership: |
|

Eloc
Genomic Map