Assay Details
| Gene Symbol: Eloc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842049.3 | XP_004842106.1 | 3 - 4 | 342 | 110 | |
| XM_021256013.1 | XP_021111672.1 | 4 - 5 | 359 | 110 | ||
| XM_021256014.1 | XP_021111673.1 | 3 - 4 | 508 | 110 | ||
| XM_021256015.1 | XP_021111674.1 | 3 - 4 | 224 | 110 | ||
More Information
Set Membership: |
|

Eloc
Genomic Map