Assay Details
| Gene Symbol: Cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842807.3 | XP_004842864.1 | 6 - 7 | 722 | 59 | |
| XM_004842808.3 | XP_004842865.1 | 6 - 7 | 743 | 59 | ||
| XM_021256676.1 | XP_021112335.1 | 6 - 7 | 561 | 59 | ||
| XM_021256677.1 | XP_021112336.1 | 6 - 7 | 634 | 59 | ||
More Information
Set Membership: |
|

Cenpn
Genomic Map