Assay Details
| Gene Symbol: LOC101712442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860252.2 | XP_004860309.1 | 13 - 14 | 1252 | 100 | |
| XM_021240737.1 | XP_021096396.1 | 13 - 14 | 1412 | 100 | ||
| XM_021240738.1 | XP_021096397.1 | 13 - 14 | 1412 | 100 | ||
More Information
Set Membership: |
|

LOC101712442
Genomic Map