Assay Details
| Gene Symbol: Sap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840197.2 | XP_004840254.1 | 1 - 2 | 202 | 107 | |
| XM_021254506.1 | XP_021110165.1 | 1 - 2 | 205 | 107 | ||
More Information
Set Membership: |
|

Sap25
Genomic Map