Assay Details
| Gene Symbol: Sap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840197.2 | XP_004840254.1 | 5 - 6 | 640 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sap25| Gene Symbol: Sap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840197.2 | XP_004840254.1 | 5 - 6 | 640 | 69 | |
Set Membership: |
|