Assay Details
Gene Symbol: Sri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004862407.3 | XP_004862464.1 | 4 - 5 | 428 | 60 | |
XM_004862408.2 | XP_004862465.1 | 4 - 5 | 285 | 60 | ||
XM_004862409.2 | XP_004862466.1 | 4 - 5 | 258 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
