Assay Details
Gene Symbol: LOC101720124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004856310.3 | XP_004856367.1 | 5 - 6 | 652 | 100 | |
XM_021237598.1 | XP_021093257.1 | 5 - 6 | 481 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
