Assay Details
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858717.3 | XP_004858774.1 | 1 - 2 | 132 | 61 | |
| XM_004858718.3 | XP_004858775.1 | 1 - 2 | 132 | 61 | ||
| XM_013077563.2 | XP_012933017.1 | 1 - 2 | 132 | 61 | ||
| XM_013077564.2 | XP_012933018.1 | 1 - 2 | 132 | 61 | ||
More Information
Set Membership: |
|

Pex19
Genomic Map