Assay Details
| Gene Symbol: Dusp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847801.2 | XP_004847858.1 | 2 - 3 | 154 | 61 | |
| XM_004847802.2 | XP_004847859.1 | 2 - 3 | 154 | 61 | ||
More Information
Set Membership: |
|

Dusp22
Genomic Map