Assay Details
Gene Symbol: Tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004869342.3 | XP_004869399.1 | 8 - 9 | 991 | 71 | |
XM_004869345.3 | XP_004869402.1 | 6 - 7 | 646 | 71 | ||
XM_004869347.3 | XP_004869404.1 | 7 - 8 | 983 | 71 | ||
XM_021248152.1 | XP_021103811.1 | 6 - 7 | 672 | 71 | ||
XM_021248153.1 | XP_021103812.1 | 9 - 10 | 1117 | 71 | ||
XM_021248154.1 | XP_021103813.1 | 6 - 7 | 1631 | 71 | ||
XM_021248155.1 | XP_021103814.1 | 5 - 6 | 616 | 71 | ||
XM_021248156.1 | XP_021103815.1 | 5 - 6 | 586 | 71 | ||
XM_021248157.1 | XP_021103816.1 | 7 - 8 | 814 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
