Assay Details
Gene Symbol: Fbxl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840382.3 | XP_004840439.1 | 6 - 7 | 2207 | 77 | |
XM_004840383.2 | XP_004840440.1 | 4 - 5 | 2101 | 77 | ||
XM_021254648.1 | XP_021110307.1 | 6 - 7 | 2249 | 77 | ||
XM_021254649.1 | XP_021110308.1 | 5 - 6 | 2186 | 77 | ||
XM_021254650.1 | XP_021110309.1 | 4 - 5 | 2038 | 77 | ||
XM_021254651.1 | XP_021110310.1 | 4 - 5 | 2037 | 77 | ||
XM_021254652.1 | XP_021110311.1 | 4 - 5 | 2052 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
