Assay Details
| Gene Symbol: Dhrs9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857621.2 | XP_004857678.1 | 2 - 3 | 471 | 65 | |
| XM_021238582.1 | XP_021094241.1 | 2 - 3 | 620 | 65 | ||
| XM_021238583.1 | XP_021094242.1 | 2 - 3 | 633 | 65 | ||
More Information
Set Membership: |
|

Dhrs9
Genomic Map