Assay Details
Gene Symbol: Wdr53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834701.3 | XP_004834758.1 | 1 - 2 | 162 | 95 | |
XM_021255041.1 | XP_021110700.1 | 1 - 2 | 162 | 95 | ||
XM_021255045.1 | XP_021110704.1 | 1 - 2 | 114 | 95 | ||
XM_021255059.1 | XP_021110718.1 | 1 - 2 | 156 | 95 | ||
XM_021255071.1 | XP_021110730.1 | 1 - 2 | 63 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004834701.3, XM_021255041.1, XM_021255045.1, XM_021255059.1, XM_021255071.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
