Assay Details
Gene Symbol: Gemin7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021250858.1 | XP_021106517.1 | 2 - 3 | 204 | 97 | |
XM_021250859.1 | XP_021106518.1 | 2 - 3 | 243 | 97 | ||
XM_021250860.1 | XP_021106519.1 | 2 - 3 | 679 | 97 | ||
XM_021250861.1 | XP_021106520.1 | 2 - 3 | 148 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021250858.1, XM_021250859.1, XM_021250860.1, XM_021250861.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
