Assay Details
Gene Symbol: Rab7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004862074.3 | XP_004862131.1 | 2 - 3 | 471 | 81 | |
XM_021242398.1 | XP_021098057.1 | 2 - 3 | 477 | 81 | ||
XM_021242399.1 | XP_021098058.1 | 3 - 4 | 571 | 81 | ||
XM_021242400.1 | XP_021098059.1 | 2 - 3 | 481 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
