Assay Details
Gene Symbol: Med19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013067483.2 | XP_012922937.1 | 1 - 2 | 455 | 80 | |
XM_021248291.1 | XP_021103950.1 | 2 - 3 | 369 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
