Assay Details
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838393.2 | XP_004838450.1 | 3 - 4 | 513 | 74 | |
XM_004838394.3 | XP_004838451.1 | 3 - 4 | 421 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
