Assay Details
| Gene Symbol: Adrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841005.3 | XP_004841062.1 | 8 - 9 | 1100 | 66 | |
| XM_004841006.3 | XP_004841063.1 | 8 - 9 | 1157 | 66 | ||
| XM_004841007.3 | XP_004841064.1 | 7 - 8 | 986 | 66 | ||
More Information
Set Membership: |
|

Adrm1
Genomic Map