Assay Details
| Gene Symbol: Fip1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849715.3 | XP_004849772.1 | 17 - 18 | 1891 | 73 | |
| XM_004849716.3 | XP_004849773.1 | 17 - 18 | 1872 | 73 | ||
| XM_004849717.3 | XP_004849774.1 | 17 - 18 | 1849 | 73 | ||
| XM_004849718.3 | XP_004849775.1 | 16 - 17 | 1743 | 73 | ||
| XM_004849719.3 | XP_004849776.1 | 15 - 16 | 1715 | 73 | ||
| XM_004849720.3 | XP_004849777.1 | 14 - 15 | 1669 | 73 | ||
| XM_013073967.2 | XP_012929421.1 | 18 - 19 | 1918 | 73 | ||
| XM_013073968.2 | XP_012929422.1 | 17 - 18 | 1812 | 73 | ||
| XM_021262342.1 | XP_021118001.1 | 18 - 19 | 1723 | 73 | ||
| XM_021262343.1 | XP_021118002.1 | 15 - 16 | 1781 | 73 | ||
More Information
Set Membership: |
|

Fip1l1
Genomic Map