Assay Details
Gene Symbol: LOC101718541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845458.3 | XP_004845515.1 | 2 - 3 | 508 | 68 | |
XM_004845460.3 | XP_004845517.1 | 2 - 3 | 229 | 68 | ||
XM_013072522.2 | XP_012927976.1 | 2 - 3 | 160 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
