Assay Details
Gene Symbol: LOC101718541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845458.3 | XP_004845515.1 | 4 - 5 | 682 | 74 | |
XM_004845460.3 | XP_004845517.1 | 4 - 5 | 403 | 74 | ||
XM_013072522.2 | XP_012927976.1 | 4 - 5 | 334 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
