Assay Details
Gene Symbol: Adgrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847140.3 | XP_004847197.2 | 4 - 5 | 733 | 79 | |
XM_021259697.1 | XP_021115356.1 | 4 - 5 | 734 | 79 | ||
XM_021259698.1 | XP_021115357.1 | 4 - 5 | 735 | 79 | ||
XM_021259699.1 | XP_021115358.1 | 4 - 5 | 734 | 79 | ||
XM_021259700.1 | XP_021115359.1 | 3 - 4 | 632 | 79 | ||
XM_021259701.1 | XP_021115360.1 | 2 - 3 | 589 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
