Assay Details
| Gene Symbol: Sae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867074.3 | XP_004867131.1 | 4 - 5 | 640 | 62 | |
| XM_004867075.3 | XP_004867132.1 | 4 - 5 | 643 | 62 | ||
| XM_013066350.2 | XP_012921804.1 | 4 - 5 | 644 | 62 | ||
More Information
Set Membership: |
|

Sae1
Genomic Map