Assay Details
| Gene Symbol: Akr1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872931.3 | XP_004872988.1 | 7 - 8 | 1014 | 65 | |
| XM_004872932.2 | XP_004872989.1 | 8 - 9 | 1085 | 65 | ||
| XM_004872933.3 | XP_004872990.1 | 6 - 7 | 894 | 65 | ||
| XM_021250775.1 | XP_021106434.1 | 7 - 8 | 969 | 65 | ||
More Information
Set Membership: |
|

Akr1a1
Genomic Map