Assay Details
| Gene Symbol: Wsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843696.3 | XP_004843753.1 | 7 - 8 | 1018 | 69 | |
| XM_013071739.2 | XP_012927193.1 | 7 - 8 | 1018 | 69 | ||
More Information
Set Membership: |
|

Wsb2
Genomic Map