Assay Details
Gene Symbol: Folr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013065041.2 | XP_012920495.1 | 5 - 6 | 708 | 61 | |
XM_013065042.2 | XP_012920496.1 | 6 - 7 | 872 | 61 | ||
XM_013065043.2 | XP_012920497.1 | 5 - 6 | 772 | 61 | ||
XM_021243338.1 | XP_021098997.1 | 4 - 5 | 665 | 61 | ||
XM_021243339.1 | XP_021098998.1 | 6 - 7 | 803 | 61 | ||
XM_021243341.1 | XP_021099000.1 | 7 - 8 | 938 | 61 | ||
XM_021243342.1 | XP_021099001.1 | 6 - 7 | 906 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
