Assay Details
| Gene Symbol: Rnf138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853109.3 | XP_004853166.1 | 2 - 3 | 478 | 86 | |
| XM_004853110.3 | XP_004853167.2 | 2 - 3 | 354 | 86 | ||
| XM_021265112.1 | XP_021120771.1 | 2 - 3 | 507 | 86 | ||
| XM_021265113.1 | XP_021120772.1 | 2 - 3 | 343 | 86 | ||
| XM_021265114.1 | XP_021120773.1 | 2 - 3 | 503 | 86 | ||
More Information
Set Membership: |
|

Rnf138
Genomic Map