Assay Details
Gene Symbol: Gpaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837758.3 | XP_004837815.1 | 1 - 2 | 213 | 59 | |
XM_004837762.3 | XP_004837819.1 | 1 - 2 | 217 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
