Assay Details
Gene Symbol: Hspbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866662.3 | XP_004866719.1 | 2 - 3 | 521 | 68 | |
XM_004866663.3 | XP_004866720.1 | 2 - 3 | 524 | 68 | ||
XM_021245938.1 | XP_021101597.1 | 2 - 3 | 394 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
