Assay Details
Gene Symbol: Coq7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855802.3 | XP_004855859.1 | 3 - 4 | 489 | 71 | |
XM_013076453.2 | XP_012931907.1 | 3 - 4 | 489 | 71 | ||
XM_021237352.1 | XP_021093011.1 | 3 - 4 | 456 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
