Assay Details
| Gene Symbol: Cutc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838649.2 | XP_004838706.1 | 8 - 9 | 820 | 66 | |
| XM_021253188.1 | XP_021108847.1 | 8 - 9 | 676 | 66 | ||
More Information
Set Membership: |
|

Cutc
Genomic Map