Assay Details
Gene Symbol: Tnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004854924.2 | XP_004854981.1 | 11 - 12 | 1104 | 66 | |
XM_021236442.1 | XP_021092101.1 | 10 - 11 | 963 | 66 | ||
XM_021236443.1 | XP_021092102.1 | 10 - 11 | 998 | 66 | ||
XM_021236444.1 | XP_021092103.1 | 11 - 12 | 1093 | 66 | ||
XM_021236445.1 | XP_021092104.1 | 10 - 11 | 998 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
