Assay Details
| Gene Symbol: Chrnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855264.3 | XP_004855321.1 | 5 - 6 | 550 | 64 | |
| XM_004855265.3 | XP_004855322.1 | 4 - 5 | 496 | 64 | ||
| XM_004855266.3 | XP_004855323.1 | 3 - 4 | 344 | 64 | ||
| XM_021236736.1 | XP_021092395.1 | 4 - 5 | 1211 | 64 | ||
More Information
Set Membership: |
|

Chrnb3
Genomic Map