Assay Details
Gene Symbol: Duoxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004835805.2 | XP_004835862.1 | 2 - 3 | 357 | 62 | |
XM_021261695.1 | XP_021117354.1 | 2 - 3 | 850 | 62 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
