Assay Details
Gene Symbol: LOC101701161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004874640.2 | XP_004874697.1 | 2 - 3 | 375 | 81 | |
XM_021252074.1 | XP_021107733.1 | 2 - 3 | 414 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
