Assay Details
Gene Symbol: Pmm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845510.3 | XP_004845567.1 | 6 - 7 | 613 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
