Assay Details
Gene Symbol: Hacd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838346.2 | XP_004838403.1 | 3 - 4 | 1773 | 83 | |
XM_013069754.2 | XP_012925208.1 | 3 - 4 | 908 | 83 | ||
XM_021252908.1 | XP_021108567.1 | 4 - 5 | 1524 | 83 | ||
XM_021252909.1 | XP_021108568.1 | 3 - 4 | 1802 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
