Assay Details
| Gene Symbol: Ensa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853985.3 | XP_004854042.1 | 3 - 4 | 477 | 55 | |
| XM_021265891.1 | XP_021121550.1 | 2 - 3 | 351 | 55 | ||
More Information
Set Membership: |
|

Ensa
Genomic Map