Assay Details
Gene Symbol: Ly6e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837856.3 | XP_004837913.1 | 3 - 4 | 364 | 78 | |
XM_021251244.1 | XP_021106903.1 | 4 - 5 | 401 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
