Assay Details
Gene Symbol: Rspo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850986.2 | XP_004851043.1 | 2 - 3 | 3687 | 55 | |
XM_021263360.1 | XP_021119019.1 | 3 - 4 | 828 | 55 | ||
XM_021263361.1 | XP_021119020.1 | 3 - 4 | 1685 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
