Assay Details
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004874125.3 | XP_004874182.1 | 2 - 3 | 222 | 66 | |
XM_004874126.3 | XP_004874183.1 | 2 - 3 | 222 | 66 | ||
XM_013069029.2 | XP_012924483.1 | 2 - 3 | 222 | 66 | ||
XM_013069030.2 | XP_012924484.1 | 2 - 3 | 222 | 66 | ||
XM_021251652.1 | XP_021107311.1 | 2 - 3 | 222 | 66 | ||
XM_021251653.1 | XP_021107312.1 | 2 - 3 | 222 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
