Assay Details
Gene Symbol: Cenpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013075340.2 | XP_012930794.1 | 2 - 3 | 356 | 75 | |
XM_021265405.1 | XP_021121064.1 | 3 - 4 | 459 | 75 | ||
XM_021265406.1 | XP_021121065.1 | 2 - 3 | 359 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
