Assay Details
| Gene Symbol: Ifnar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842303.3 | XP_004842360.1 | 3 - 4 | 1284 | 75 | |
| XM_004842305.3 | XP_004842362.1 | 5 - 6 | 561 | 75 | ||
| XM_013071305.2 | XP_012926759.1 | 3 - 4 | 395 | 75 | ||
| XM_021256192.1 | XP_021111851.1 | 4 - 5 | 481 | 75 | ||
| XM_021256193.1 | XP_021111852.1 | 5 - 6 | 2450 | 75 | ||
More Information
Set Membership: |
|

Ifnar2
Genomic Map