Assay Details
Gene Symbol: Rangrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004857491.3 | XP_004857548.1 | 2 - 3 | 305 | 88 | |
XM_013076842.2 | XP_012932296.1 | 2 - 3 | 305 | 88 | ||
XM_021238502.1 | XP_021094161.1 | 2 - 3 | 257 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
