Assay Details
| Gene Symbol: LOC101700516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844346.2 | XP_004844403.1 | 7 - 8 | 1084 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101700516| Gene Symbol: LOC101700516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844346.2 | XP_004844403.1 | 7 - 8 | 1084 | 82 | |
Set Membership: |
|