Assay Details
| Gene Symbol: Khdrbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837910.2 | XP_004837967.1 | 8 - 9 | 1019 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khdrbs3| Gene Symbol: Khdrbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837910.2 | XP_004837967.1 | 8 - 9 | 1019 | 67 | |
Set Membership: |
|