Assay Details
| Gene Symbol: Tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840086.2 | XP_004840143.1 | 5 - 6 | 600 | 63 | |
| XM_021254433.1 | XP_021110092.1 | 5 - 6 | 602 | 63 | ||
| XM_021254434.1 | XP_021110093.1 | 4 - 5 | 408 | 63 | ||
More Information
Set Membership: |
|

Tmem120a
Genomic Map