Assay Details
Gene Symbol: Snrpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852670.3 | XP_004852727.1 | 1 - 2 | 186 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
